Ethyl isoxazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A866471-1G A866471-25G A866471-5G
76807-754EA 32 USD
76807-754 76807-756 77670-254
Ethyl isoxazole-3-carboxylate 97%
Ethyl isoxazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₃
MDL Number: MFCD08234628
CAS Number: 3209-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR