Ethyl isoxazole-4-carboxylate 98%

Supplier: Ambeed
Warning

A864326-1G A864326-250MG A864326-5G
76807-724EA 167.63 USD
76807-724 76807-726 76807-728
Ethyl isoxazole-4-carboxylate 98%
Ethyl isoxazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₃
MDL Number: MFCD07368244
CAS Number: 80370-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR