2-((4-((7-Chloroquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol 98%

Supplier: Ambeed
Warning

A862774-100G A862774-1G A862774-250MG A862774-5G
76807-714EA 567 USD
76807-714 76807-716 76807-718 76807-720
2-((4-((7-Chloroquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol 98%
2-((4-((7-Chloroquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol

Precaución: for Research Use Only

Formula: C₁₈H₂₆ClN₃O
MDL Number: MFCD00242707
CAS Number: 118-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR