Lithium dihydrogen orthophosphate 99%

Supplier: Ambeed
Warning

Synonyms: Lithium dihydrogen phosphate, Lithium phosphate monobasic, Lithium dihydrogenorthophosphate, Lithium dihydrogenphosphate

A850874-1000G A850874-100G A850874-25G A850874-500G
76807-690EA 101 USD
76807-690 76807-692 76807-694 76807-696
Lithium dihydrogen orthophosphate 99%
Lithium dihydrogen orthophosphate

Precaución: for Research Use Only

Formula: H₂LiO₄P
MW: 103.93
MDL Number: MFCD00016188
CAS Number: 13453-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR