Ethyl-4,6-dichloroquinoline-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4,6-dichloroquinoline-3-carboxylate

A847227-100MG A847227-1G A847227-250MG A847227-5G
76807-678EA 20.16 USD
76807-678 76807-680 76807-682 76807-684
Ethyl-4,6-dichloroquinoline-3-carboxylate 95%
Ethyl-4,6-dichloroquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₉Cl₂NO₂
MW: 270.109 g/mol
MDL Number: MFCD00173348
CAS Number: 21168-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR