Imidazo[1,2-a]pyridine-7-carboxylic acid 95%

Supplier: Ambeed
Warning

A844394-100MG A844394-1G A844394-250MG A844394-5G
76807-670EA 13 USD
76807-670 77573-684 77880-644 77880-645
Imidazo[1,2-a]pyridine-7-carboxylic acid 95%
Imidazo[1,2-a]pyridine-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD09834781
CAS Number: 648423-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR