Dimethyl 2,2'-disulfanediyldibenzoate 96%

Supplier: Ambeed
Warning

A844227-1G A844227-5G
76807-668EA 44 USD
76807-668 77880-643
Dimethyl 2,2'-disulfanediyldibenzoate 96%
Dimethyl 2,2'-disulfanediyldibenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₄S₂
MDL Number: MFCD00025860
CAS Number: 5459-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR