α-Linolenic acid 97% (by GC)

Supplier: Ambeed
Warning

Synonyms: ALA, (9Z,12Z,15Z)-Octadeca-9,12,15-trienoic acid, Linolenic acid, 9Z,12Z,15Z-Octadecatrienoic acid, cis,cis,cis-9,12,15-Octadecatrienoic acid

A843943-250MG A843943-25G A843943-1G
76807-664EA 11.67 USD
76807-664 76807-666 77552-578
α-Linolenic acid 97% (by GC)
α-Linolenic acid

Precaución: for Research Use Only

Formula: C₁₈H₃₀O₂
MW: 278.44 g/mol
MDL Number: MFCD00065720
CAS Number: 463-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR