2,3-Dichloromaleic anhydride 97%

Supplier: Ambeed
Warning

A842166-1G A842166-25G A842166-5G
76807-660EA 8 USD
76807-660 77573-678 77595-346
2,3-Dichloromaleic anhydride 97%
2,3-Dichloromaleic anhydride

Precaución: for Research Use Only

Formula: C₄Cl₂O₃
MDL Number: MFCD00005520
CAS Number: 1122-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR