Ethylenediaminetetraacetic acid manganese disodium salt hydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
EDTA disodium manganese, Manganese Disodium Ethylenediaminetetraacetate, Manganese(II) disodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate
A840775-1000G
A840775-100G
A840775-500G
76807-656EA
20
USD
76807-656
77603-138
77603-140
Ethylenediaminetetraacetic acid manganese disodium salt hydrate 98%
Ethylenediaminetetraacetic acid manganese disodium salt hydrate
Precaución: for Research Use Only
Formula:
C₁₀H₁₆N₂O₈ MW: 389.13 g/mol |
MDL Number:
MFCD00054451 CAS Number: 15375-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...