Dimethyl sebacate 98%

Supplier: Ambeed
Danger

Synonyms: 1,10-Decanedioic acid dimethyl ester, Dimethyl decanedioate, Sebacic acid dimethyl ester

A836928-25G A836928-100G
76807-654EA 6 USD
76807-654 77603-126
Dimethyl sebacate 98%
Dimethyl sebacate

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₄
MW: 230.3 g/mol
Boiling Pt: 102
Melting Pt: 38
MDL Number: MFCD00008472
CAS Number: 106-79-6
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR