DL-2-Hydroxybutyric acid 95%

Supplier: Ambeed
Danger

A836049-1G A836049-250MG
76807-650EA 64 USD
76807-650 76807-652
DL-2-Hydroxybutyric acid 95%
DL-2-Hydroxybutyric acid

Precaución: for Research Use Only

Formula: C₄H₈O₃
MDL Number: MFCD00070502
CAS Number: 600-15-7
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR