Dodecylamine acetate 97%

Supplier: Ambeed
Warning

Synonyms: Dodecylamineacetate, Aminododecane Acetate, Laurylamine Acetate

A829166-1G A829166-25G A829166-5G
76807-628EA 13.8 USD
76807-628 76807-630 76807-632
Dodecylamine acetate 97%
Dodecylamine acetate

Precaución: for Research Use Only

Formula: C₁₂H₂₇N
MW: 245.41 g/mol
MDL Number: MFCD00043524
CAS Number: 2016-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR