4-Aminoantipyrine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-AA.HCl, ['4-AA·HCl'], 4-Aminoantipyrine Hydrochloride, 4-Amino-1,5-dimethyl-2-phenyl-1H-pyrazol-3(2H)-one hydrochloride

A824260-1G A824260-25G A824260-5G
76807-622EA 5 USD
76807-622 76807-624 76807-626
4-Aminoantipyrine hydrochloride 98%
4-Aminoantipyrine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃O
MW: 239.70 g/mol
MDL Number: MFCD00042012
CAS Number: 22198-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR