Diphenyl(1,5-cyclooctadiene)platinum(II) 98%

Supplier: Ambeed
Warning

Synonyms: (1,5-Cyclooctadiene)diphenylplatinum(II)

A822885-100MG A822885-1G
76807-618EA 58.35 USD
76807-618 76807-620
Diphenyl(1,5-cyclooctadiene)platinum(II) 98%
Diphenyl(1,5-cyclooctadiene)platinum(II)

Precaución: for Research Use Only

Formula: C₂₀H₂₂Pt
MW: 457.47 g/mol
MDL Number: MFCD03788255
CAS Number: 12277-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR