4-(4-Biphenylyl)-4-oxobutyric acid 99%

Supplier: Ambeed
Warning

Synonyms: Fenbufen, 4-([1,1'-Biphenyl]-4-yl)-4-oxobutanoic acid, Cinopal, Napanol, Lederfen, Bufemid

A819665-100MG A819665-25G A819665-5G A819665-100G A819665-1000G A819665-500G
76807-598EA 5 USD
76807-598 76807-600 76807-602 77603-088 77670-160 77670-162
4-(4-Biphenylyl)-4-oxobutyric acid 99%
4-(4-Biphenylyl)-4-oxobutyric acid

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₃
MW: 254.285 g/mol
MDL Number: MFCD00056701
CAS Number: 36330-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR