1-(Toluene-4-sulfonyl)piperazine 95%

Supplier: Ambeed
Warning

A818471-1G A818471-25G A818471-5G
76807-592EA 19 USD
76807-592 76807-594 76807-596
1-(Toluene-4-sulfonyl)piperazine 95%
1-(Toluene-4-sulfonyl)piperazine
Formula: C₁₁H₁₆N₂O₂S
MDL Number: MFCD02018961
CAS Number: 27106-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR