(R)-1-Benzyl-3-pyrrolidinol 95%

Supplier: Ambeed
Warning

Synonyms: (R)-1-Benzyl-3-hydroxypyrrolidine, (R)-(+)-1-Benzyl-3-pyrrolidinol, (R)-3-Hydroxy-1-benzylpyrrolidine

A818005-1G A818005-25G A818005-5G
76807-586EA 12 USD
76807-586 76807-588 76807-590
(R)-1-Benzyl-3-pyrrolidinol 95%
(R)-1-Benzyl-3-pyrrolidinol

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO
MDL Number: MFCD00075484
CAS Number: 101930-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR