Sodium isoascorbate monohydrate 99%

Supplier: Ambeed
Warning

Synonyms: Isoascorbic acid Sodium salt, Sodium Erythorbate, Sodium (R)-2-((R)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Erythorbic acid Sodium salt, Sodium D-Araboascorbate, D-Araboascorbic acid Sodium salt

A811818-1000G A811818-100G A811818-25G A811818-500G
76807-572EA 40.32 USD
76807-572 76807-574 76807-576 76807-578
Sodium isoascorbate monohydrate 99%
Sodium isoascorbate monohydrate

Precaución: for Research Use Only

Formula: C₆H₈O₆
MW: 198.11 g/mol
MDL Number: MFCD00067298
CAS Number: 6381-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR