4-Methyl-N-tosylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: Bis(p-tolylsulphonyl)amine, Ditosylamine, Bis(p-toluenesulphonyl)amine, Bis(4-methylbenzenesulphonyl)amine

A808528-1G A808528-250MG
76807-554EA 50 USD
76807-554 77880-607
4-Methyl-N-tosylbenzenesulfonamide 98%
4-Methyl-N-tosylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₄S₂
MDL Number: MFCD00014917
CAS Number: 3695-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR