Ethyl-1H-1,2,4-triazol-1-ylacetate 97%

Supplier: Ambeed
Warning

A807326-1G A807326-250MG
76807-552EA 351 USD
76807-552 77880-606
Ethyl-1H-1,2,4-triazol-1-ylacetate 97%
Ethyl-1H-1,2,4-triazol-1-ylacetate

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MDL Number: MFCD00215199
CAS Number: 56563-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR