Sodium-DL-3-hydroxybutyrate 98%

Supplier: Ambeed
Warning

A806846-1000G A806846-100G A806846-10G A806846-25G A806846-500G A806846-5G
76807-540EA 123 USD
76807-540 76807-542 76807-544 76807-546 76807-548 76807-550
Sodium-DL-3-hydroxybutyrate 98%
Sodium-DL-3-hydroxybutyrate

Precaución: for Research Use Only

Formula: C₄H₈O₃
MDL Number: MFCD00016716
CAS Number: 150-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR