Diethanolamine hydrochloride 95%

Supplier: Ambeed
Warning

A806064-100G A806064-25G A806064-5G
76807-534EA 68 USD
76807-534 76807-536 76807-538
Diethanolamine hydrochloride 95%
Diethanolamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁NO₂
MDL Number: MFCD00012595
CAS Number: 14426-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR