Isobutyl cinnamate 98%

Supplier: Ambeed
Warning

A802989-100G A802989-25G A802989-500G
76807-518EA 23.34 USD
76807-518 76807-520 76807-522
Isobutyl cinnamate 98%
Isobutyl cinnamate

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
MDL Number: MFCD00038289
CAS Number: 122-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR