Ethylene glycol dibutyl ether 98%

Supplier: Ambeed
Warning

Synonyms: Ethyleneglycoldi-n-butylether, Dibutyl Glycol, Dibutyl Cellosolve, 1,2-Dibutoxyethane

A799476-100G A799476-25G A799476-500G
76807-512EA 42.44 USD
76807-512 76807-514 76807-516
Ethylene glycol dibutyl ether 98%
Ethylene glycol dibutyl ether

Precaución: for Research Use Only

Formula: C₁₀H₂₂O₂
MW: 174.28 g/mol
MDL Number: MFCD00048785
CAS Number: 112-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR