Ethyl-2,4-dioxo-4-(2-pyridinyl)butanoate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2,4-dioxo-4-(pyridin-2-yl)butanoate

A799034-1G A799034-250MG A799034-5G
76807-506EA 70.02 USD
76807-506 76807-508 76807-510
Ethyl-2,4-dioxo-4-(2-pyridinyl)butanoate 95%
Ethyl-2,4-dioxo-4-(2-pyridinyl)butanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₄
MW: 221.21 g/mol
MDL Number: MFCD08056636
CAS Number: 92288-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR