ε-Caprolactam 99%

Supplier: Ambeed
Warning

Synonyms: Azepan-2-one, 2-Oxohexamethyleneimine, 6-Aminocaproic acid lactam

A798855-1000G A798855-100G A798855-25G A798855-500G
76807-498EA 18.04 USD
76807-498 76807-500 76807-502 76807-504
ε-Caprolactam 99%
ε-Caprolactam

Precaución: for Research Use Only

Formula: C₆H₁₁NO
MW: 113.16 g/mol
MDL Number: MFCD00006936
CAS Number: 105-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR