Hexyl-4-hydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxybenzoic acid hexyl ester, Hexyl 4-hydroxybenzoate, Hexylparaben

A798570-100G A798570-25G A798570-5G
76807-492EA 171.87 USD
76807-492 76807-494 76807-496
Hexyl-4-hydroxybenzoate 98%
Hexyl-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
MDL Number: MFCD00016480
CAS Number: 1083-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR