Sodium diformylamide 90%

Supplier: Ambeed
Warning

Synonyms: Sodium diformamide, Sodium Diformylamide, Diformylamide Sodium salt

A798151-100G A798151-10G A798151-1G A798151-25G A798151-5G
76807-482EA 90 USD
76807-482 76807-484 76807-486 76807-488 76807-490
Sodium diformylamide 90%
Sodium diformylamide

Precaución: for Research Use Only

Formula: C₂H₃NNaO₂
MDL Number: MFCD00145302
CAS Number: 18197-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR