1-(4-Nitrobenzyl)-1H-imidazole 98% (contains < 10% H₂O)

Supplier: Ambeed
Warning

A796230-1G A796230-250MG A796230-25G A796230-5G
76807-464EA 14 USD
76807-464 76807-466 76807-468 76807-470
1-(4-Nitrobenzyl)-1H-imidazole 98% (contains < 10% H₂O)
1-(4-Nitrobenzyl)-1H-imidazole

Precaución: for Research Use Only

Formula: C₁₀H₉N₃O₂
MDL Number: MFCD01799096
CAS Number: 18994-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR