Dihexyl phthalate 98%

Supplier: Ambeed
Danger

A794208-10G A794208-1G A794208-25G A794208-5G A794208-100G
76807-452EA 65.78 USD
76807-452 76807-454 76807-456 76807-458 77627-584
Dihexyl phthalate 98%
Dihexyl phthalate

Precaución: for Research Use Only

Formula: C₂₀H₃₀O₄
MDL Number: MFCD00043720
CAS Number: 84-75-3
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR