H-Hyp-OBzl·HCl 98%

Supplier: Ambeed
Warning

Synonyms: H-Hyp-OBzl·HCl, Benzyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate hydrochloride

A793897-1G A793897-5G
76807-448EA 6.37 USD
76807-448 76807-450
H-Hyp-OBzl·HCl 98%
H-Hyp-OBzl·HCl

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃·HCl
MW: 257.714 g/mol
MDL Number: MFCD00237998
CAS Number: 62147-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR