Ethyl phenoxyacetate 98%

Supplier: Ambeed
Warning

A793732-100G A793732-1G A793732-25G A793732-5G
76807-440EA 60 USD
76807-440 76807-442 76807-444 76807-446
Ethyl phenoxyacetate 98%
Ethyl phenoxyacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00026895
CAS Number: 2555-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR