1,2-Distearoyl-sn-glycero-3-phosphocholine 99%

Supplier: Ambeed
Warning

Synonyms: DSPC, 1,2-Distearoyl-sn-glycero-3-phosphorylcholine, 1,2-Dioctadecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(stearoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, (R)-2,3-Bis(stearoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate

A793066-1G A793066-5G A793066-250MG A793066-25G
76807-438EA 77.45 USD
76807-438 77573-538 77603-026 77670-118
1,2-Distearoyl-sn-glycero-3-phosphocholine 99%
1,2-Distearoyl-sn-glycero-3-phosphocholine

Precaución: for Research Use Only

Formula: C₄₄H₈₈NO₈P
MW: 790.16 g/mol
MDL Number: MFCD00036905
CAS Number: 816-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR