Dimethyl(2-oxoheptyl)phosphonate ≥95%

Supplier: Ambeed
Warning

A793019-1G A793019-250MG A793019-5G
76807-432EA 23 USD
76807-432 76807-434 76807-436
Dimethyl(2-oxoheptyl)phosphonate ≥95%
Dimethyl(2-oxoheptyl)phosphonate

Precaución: for Research Use Only

Formula: C₉H₁₉O₄P
MDL Number: MFCD00009514
CAS Number: 36969-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR