Glycyl-DL-leucine 98+%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Aminoacetamido)-4-methylpentanoic acid, H-Gly-DL-Leu-OH

A787890-100G A787890-1G A787890-25G A787890-5G
76807-420EA 519.85 USD
76807-420 76807-422 76807-424 76807-426
Glycyl-DL-leucine 98+%
Glycyl-DL-leucine

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₃
MW: 188.23 g/mol
MDL Number: MFCD00066051
CAS Number: 688-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR