N-(4-Aminophenyl)maleimide 93%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Aminophenyl)-1H-pyrrole-2,5-dione

A785555-100MG A785555-1G A785555-250MG
76807-414EA 55.17 USD
76807-414 76807-416 76807-418
N-(4-Aminophenyl)maleimide 93%
N-(4-Aminophenyl)maleimide

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
MDL Number: MFCD00051191
CAS Number: 29753-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR