(S)-(+)-Ethyl mandelate 97%

Supplier: Ambeed
Warning

Synonyms: L-(+)-Mandelic acid ethyl ester, (S)-Ethyl 2-hydroxy-2-phenylacetate, Ethyl L-(+)-Mandelate

A784137-100G A784137-1G A784137-25G A784137-5G
76807-406EA 93 USD
76807-406 76807-408 76807-410 76807-412
(S)-(+)-Ethyl mandelate 97%
(S)-(+)-Ethyl mandelate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00064249
CAS Number: 13704-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR