Indium(III) hydroxide 99+%

Supplier: Ambeed
Danger

A781222-100G A781222-25G A781222-5G
76807-382EA 135 USD
76807-382 76807-384 76807-386
Indium(III) hydroxide 99+%
Indium(III) hydroxide

Precaución: for Research Use Only

Formula: H₃InO₃
MW: 165.84 g/mol
MDL Number: MFCD00011056
CAS Number: 20661-21-6
UN: 3288
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR