Gold(I) iodide 99+%
Supplier: Ambeed
|
Warning
|
A780784-100MG
A780784-1G
A780784-250MG
76807-376EA
83
USD
76807-376
76807-378
76807-380
Gold(I) iodide 99+%
Gold(I) iodide
Precaución: for Research Use Only
Formula:
AuI MW: 323.87 g/mol |
MDL Number:
MFCD00014175 CAS Number: 10294-31-2 Merck Index: 12,04530 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...