Methacrylamide 98%

Supplier: Ambeed
Warning

A780121-1000G A780121-100G A780121-25G A780121-500G
76807-368EA 46.68 USD
76807-368 76807-370 76807-372 76807-374
Methacrylamide 98%
Methacrylamide

Precaución: for Research Use Only

Formula: C₄H₇NO
MW: 85.11 g/mol
MDL Number: MFCD00008018
CAS Number: 79-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR