Ethyl-2-chloro-4-methyl-1,3-thiazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-chloro-4-methylthiazole-5-carboxylate

A778736-10G A778736-1G A778736-5G
76807-362EA 176.11 USD
76807-362 76807-364 76807-366
Ethyl-2-chloro-4-methyl-1,3-thiazole-5-carboxylate 98%
Ethyl-2-chloro-4-methyl-1,3-thiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈ClNO₂S
MW: 205.66 g/mol
MDL Number: MFCD02956798
CAS Number: 7238-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR