Fmoc-L-Ser(α-D-GalNAc(Ac)3)-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-L-Ser (α-D-GalNAc(Ac)3)-OH, N-α-Fmoc-O-β-(2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-galactopyranosyl)-L-serine, Fmoc-Ser(GalNAc(Ac)₃-α-D)-OH, Fmoc-Ser(O-α-D-GalNAc(OAc)3)-OH, Fmoc-ser(galnac(ac)3-alpha-d)-OH
A772566-10MG
A772566-1MG
A772566-25MG
A772566-5MG
76807-322EA
80
USD
76807-322
76807-324
76807-326
76807-328
Fmoc-L-Ser(α-D-GalNAc(Ac)3)-OH 97%
Fmoc-L-Ser(alpha-D-GalNAc(Ac)3)-OH
Precaución: for Research Use Only
Formula:
C₃₂H₃₆N₂O₁₃ |
MDL Number:
MFCD00171375 CAS Number: 120173-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...