3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one 98%

Supplier: Ambeed
Warning

A771581-100G A771581-10G A771581-1G A771581-25G A771581-5G
76807-300EA 104 USD
76807-300 76807-302 76807-304 76807-306 76807-308
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one 98%
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₃
MDL Number: MFCD00006652
CAS Number: 69225-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR