DL-Citrulline 97%

Supplier: Ambeed
Warning

Synonyms: H-DL-Cit-OH

A769535-100G A769535-100MG A769535-5G A769535-25G A769535-1G A769535-50MG
76807-292EA 481 USD
76807-292 76807-294 76807-296 77308-292 77573-482 77627-532
DL-Citrulline 97%
DL-Citrulline

Precaución: for Research Use Only

Formula: C₆H₁₃N₃O₃
MDL Number: MFCD00007955
CAS Number: 627-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR