N-(tert-Butoxycarbonyl)-4-phenyl-L-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-4-phenyl-L-phenylalanine, Boc-Bip(4,4')-OH, Boc-4-phenyl-Phe-OH, Boc-Bip-OH

A768159-10G A768159-1G A768159-250MG A768159-5G A768159-25G
76807-284EA 180.36 USD
76807-284 76807-286 76807-288 76807-290 77602-962
N-(tert-Butoxycarbonyl)-4-phenyl-L-phenylalanine 98%
N-(tert-Butoxycarbonyl)-4-phenyl-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₀H₂₃NO₄
MW: 341.41 g/mol
MDL Number: MFCD00191184
CAS Number: 147923-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR