Dimethyl adipate 99%

Supplier: Ambeed
Warning

Synonyms: Adipic acid dimethyl ester, 1,6-Hexanedioic acid dimethyl ester

A764010-1000G A764010-100G A764010-25G A764010-500G
76807-266EA 23 USD
76807-266 76807-268 76807-270 76807-272
Dimethyl adipate 99%
Dimethyl adipate

Precaución: for Research Use Only

Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 229
Melting Pt: 10.3
Flash Pt: 107
MDL Number: MFCD00008469
CAS Number: 627-93-0
Merck Index: 13,00163

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR