tert-Butyl 2-carbamoylpiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butoxycarbonyl)piperidine-2-carboxamide

A762361-10G A762361-1G A762361-25G A762361-5G
76807-258EA 72.15 USD
76807-258 76807-260 76807-262 76807-264
tert-Butyl 2-carbamoylpiperidine-1-carboxylate 98%
tert-Butyl 2-carbamoylpiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₃
MW: 228.29 g/mol
MDL Number: MFCD02183582
CAS Number: 388077-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR