Lacidipine 98%

Supplier: Ambeed
Warning

Synonyms: (E)-Diethyl 4-(2-(3-(tert-butoxy)-3-oxoprop-1-en-1-yl)phenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate

A761993-100G A761993-1G A761993-50MG A761993-5G
76807-250EA 388.29 USD
76807-250 76807-252 76807-254 76807-256
Lacidipine 98%
Lacidipine

Precaución: for Research Use Only

Formula: C₂₆H₃₃NO₆
MW: 455.55 g/mol
MDL Number: MFCD00865936
CAS Number: 103890-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR