(S)-1-Benzyl 3-ethyl piperidine-1,3-dicarboxylate 97%

Supplier: Ambeed
Warning

A761864-1G A761864-250MG A761864-5G
76807-244EA 55.17 USD
76807-244 76807-246 76807-248
(S)-1-Benzyl 3-ethyl piperidine-1,3-dicarboxylate 97%
(S)-1-Benzyl 3-ethyl piperidine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₄
MW: 291.34 g/mol
MDL Number: MFCD07374392
CAS Number: 174699-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR